Please login to your online account to display your discounted pricing

anti-RGS13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224415SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224415SS

  • / Each
 

Description & Specifications

Specifications

Antigen RGS13
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation 100μL of crude rabbit sera with 50% glycerol
Gene Accession No. O14921
Host Species Rabbit
Immunogen Within the range of amino acids 111-139 of human RGS13 protein were used as the immunogen.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6003
Target Species Human