Please login to your online account to display your discounted pricing

anti-RGS13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224415

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224415

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL of crude rabbit sera with 50% glycerol
Gene Accession No. O14921
Gene ID (Entrez) 6003
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Antigen RGS13
Immunogen Within the range of amino acids 111-139 of human RGS13 protein were used as the immunogen.
Primary or Secondary Primary
Format Antiserum
Applications Western Blotting
Quantity 0.1mL
Cross Reactivity Human
Host Species Rabbit