Please login to your online account to display your discounted pricing

anti-RGS11, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 201-217 (SADFHKREIEYFRKALG) of human RGS11.
Antigen RGS11
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056658

 
novus biologicals canada
NB10056658
Unlabeled 0.2mL Each for $255.80

NB10056658T

 
novus biologicals canada
NB10056658SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 201-217 (SADFHKREIEYFRKALG) of human RGS11.
Antigen RGS11
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity An approx. 48 kDa band is observed.
Gene ID (Entrez) 8786
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL