missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-RGS1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within amino acid residues 1-50 of human RGS1 was used as immunogen.
Antigen RGS1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056737 Novus Biologicals Canada
NB10056737
0.1mg Each for $255.80
NB10056737T Novus Biologicals Canada
NB10056737SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
RGS1
Purified
RUO
Reacts with Human. 82% sequence identity with mouse and rat proteins.
5996
0.5mg/mL
A synthetic peptide within amino acid residues 1-50 of human RGS1 was used as immunogen.
Polyclonal
Primary
Western Blot
Q08116
PBS containing 0.05% BSA