Please login to your online account to display your discounted pricing

anti-RGS1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056737SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056737T

  • / Each
 

Description & Specifications

Specifications

Test Specificity Reacts with Human. 82% sequence identity with mouse and rat proteins.
Formulation PBS containing 0.05% BSA
Conjugate Unlabeled
Gene ID (Entrez) 5996
Gene Accession No. Q08116
Concentration 0.5mg/mL
Antigen RGS1
Regulatory Status RUO
Format Purified
Immunogen A synthetic peptide within amino acid residues 1-50 of human RGS1 was used as immunogen.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Host Species Rabbit
Quantity 0.025mg
Applications Western Blotting