Please login to your online account to display your discounted pricing

anti-RGS1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056737

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056737

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5996
Primary or Secondary Primary
Test Specificity Reacts with Human. 82% sequence identity with mouse and rat proteins.
Gene Accession No. Q08116
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Regulatory Status RUO
Concentration 0.5mg/mL
Immunogen A synthetic peptide within amino acid residues 1-50 of human RGS1 was used as immunogen.
Cross Reactivity Human
Host Species Rabbit
Antigen RGS1
Quantity 0.1mg
Format Purified
Applications Western Blotting