Please login to your online account to display your discounted pricing

anti-RFXANK, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Peptide made to a portion of the RFX-B protein.
Antigen RFXANK
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600205SS

 
novus biologicals canada
NB600205SS
Unlabeled 0.025mL Each for $109.00

NB600205

 
novus biologicals canada
NB600205
Unlabeled 0.1mL Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Peptide made to a portion of the RFX-B protein.
Antigen RFXANK
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Reacts with residues 18-31 [ASELGDPEDPGEEAC] of the RFX-B protein. Human and Primates RFX-B.
Gene ID (Entrez) 8625
Formulation PBS
Concentration 1mg/mL