Please login to your online account to display your discounted pricing

anti-RFXANK, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600205SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600205SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Formulation PBS
Test Specificity Reacts with residues 18-31 [ASELGDPEDPGEEAC] of the RFX-B protein. Human and Primates RFX-B.
Concentration 1mg/mL
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Primate
Antigen RFXANK
Primary or Secondary Primary
Gene ID (Entrez) 8625
Host Species Rabbit
Quantity 0.025mL
Immunogen Peptide made to a portion of the RFX-B protein.
Applications Western Blotting
Format Affinity Purified