Please login to your online account to display your discounted pricing

anti-Relaxin R1/LGR7, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human LGR7 was used as the immunogen for this antibody.
Antigen Relaxin R1/LGR7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224540

 
novus biologicals canada
NBP224540
Unlabeled 0.1mg Each for $329.00

NBP224540SS

 
novus biologicals canada
NBP224540SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human LGR7 was used as the immunogen for this antibody.
Antigen Relaxin R1/LGR7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this phospho antibody is 100% homologous in human, mouse, dog, opossum and chimpanzee.
Gene ID (Entrez) 59350
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL