Please login to your online account to display your discounted pricing

anti-Relaxin R1/LGR7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224540SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224540SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL
Conjugate Unlabeled
Gene ID (Entrez) 59350
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Antigen Relaxin R1/LGR7
Applications Western Blotting
Immunogen A portion of amino acids 50-100 of human LGR7 was used as the immunogen for this antibody.
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen for this phospho antibody is 100% homologous in human, mouse, dog, opossum and chimpanzee.
Quantity 0.025mg
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Regulatory Status RUO