Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 538-546 (DEDFSSIADM) of human NF-kB p65.
Antigen RelA/NFkB p65
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056647

 
novus biologicals canada
NB10056647
Unlabeled 0.2mL Each for $259.02

NB10056647T

 
novus biologicals canada
NB10056647SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 538-546 (DEDFSSIADM) of human NF-kB p65.
Antigen RelA/NFkB p65
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blot
Test Specificity Cross reacts with Human, Mouse and Rat. Predicted to react with New World Monkey, Pig, Dog, Avian and Gorilla.
Gene Accession No. Q969V5
Gene ID (Entrez) 5970
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL