Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Polyclonal, Novus Biologicals™

$144.63 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human NFkB p65 protein (between residues 300-400) [UniProt Q04206]
Antigen RelA/NFkB p65
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP196139

 
novus biologicals canada
NBP196139
Unlabeled 0.1mL Each for $309.05

NBP196139SS

 
novus biologicals canada
NBP196139SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human NFkB p65 protein (between residues 300-400) [UniProt Q04206]
Antigen RelA/NFkB p65
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. Q04206
Gene ID (Entrez) 5970
Formulation PBS, 30% glycerol
Concentration 1.02mg/mL