Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056647SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056647T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q969V5
Applications Simple Western
Applications Western Blotting
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 538-546 (DEDFSSIADM) of human NF-kB p65.
Quantity 0.05mL
Gene ID (Entrez) 5970
Concentration 0.5mg/mL
Conjugate Unlabeled
Antigen RelA/NFkB p65
Regulatory Status RUO
Test Specificity Cross reacts with Human, Mouse and Rat. Predicted to react with New World Monkey, Pig, Dog, Avian and Gorilla.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit