Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056647

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056647

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. Q969V5
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Concentration 0.5mg/mL
Conjugate Unlabeled
Host Species Rabbit
Quantity 0.2mL
Monoclonal or Polyclonal Polyclonal
Antigen RelA/NFkB p65
Gene ID (Entrez) 5970
Test Specificity Cross reacts with Human, Mouse and Rat. Predicted to react with New World Monkey, Pig, Dog, Avian and Gorilla.
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 538-546 (DEDFSSIADM) of human NF-kB p65.