Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP196139SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP196139SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q04206
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Concentration 1.02mg/mL
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5970
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Regulatory Status RUO
Antigen RelA/NFkB p65
Format Affinity Purified
Immunogen A synthetic peptide made to an internal portion of the human NFkB p65 protein (between residues 300-400) [UniProt Q04206]
Host Species Rabbit