Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP196138SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP196138SS

  • / Each
 

Description & Specifications

Specifications

Antigen RelA/NFkB p65
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Concentration 1.02mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q04206
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human NFkB p65 protein (between residues 500-551) [UniProt Q04206]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5970
Target Species Human
Target Species Murine
Target Species Primate