Please login to your online account to display your discounted pricing

anti-RelA/NFkB p65, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP196138

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP196138

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 1.02mg/mL
Formulation PBS, 30% glycerol
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Host Species Rabbit
Quantity 0.1mL
Immunogen A synthetic peptide made to a C-terminal portion of the human NFkB p65 protein (between residues 500-551) [UniProt Q04206]
Gene ID (Entrez) 5970
Gene Accession No. Q04206
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Regulatory Status RUO
Format Affinity Purified
Antigen RelA/NFkB p65