Please login to your online account to display your discounted pricing

anti-Reg3A, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human REG3A was used as the immunogen for this antibody.
Antigen Reg3A
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227297

 
novus biologicals canada
NBP227297
Unlabeled 0.1mg Each for $257.41

NBP227297SS

 
novus biologicals canada
NBP227297SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human REG3A was used as the immunogen for this antibody.
Antigen Reg3A
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 88% in mouse and 79% homologous in mouse.
Gene ID (Entrez) 5068
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL