Please login to your online account to display your discounted pricing

anti-Reg3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227297SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227297SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5068
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Quantity 0.025mg
Antigen Reg3A
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 88% in mouse and 79% homologous in mouse.
Primary or Secondary Primary
Immunogen A portion of amino acids 100-150 of human REG3A was used as the immunogen for this antibody.
Host Species Rabbit
Cross Reactivity Human