Please login to your online account to display your discounted pricing

anti-RCAN1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human RCAN1 (within residues 210-252). [Swiss-Prot No. P53805]
Antigen RCAN1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP146853

 
novus biologicals canada
NBP146853
Unlabeled 0.1mL Each for $309.05

NBP146853SS

 
novus biologicals canada
NBP146853SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human RCAN1 (within residues 210-252). [Swiss-Prot No. P53805]
Antigen RCAN1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human and mouse. Immunogen has 92% identity to rat.
Gene Accession No. P53805
Gene ID (Entrez) 1827
Formulation PBS, 30% glycerol
Concentration 1mg/mL