Please login to your online account to display your discounted pricing

anti-RCAN1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP146853SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP146853SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 1827
Concentration 1mg/mL
Test Specificity Human and mouse. Immunogen has 92% identity to rat.
Conjugate Unlabeled
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mL
Cross Reactivity Human
Cross Reactivity Mouse
Antigen RCAN1
Formulation PBS, 30% glycerol
Primary or Secondary Primary
Format Affinity Purified
Gene Accession No. P53805
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human RCAN1 (within residues 210-252). [Swiss-Prot No. P53805]
Regulatory Status RUO