Please login to your online account to display your discounted pricing

anti-RCAN1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP146852SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP146852SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Test Specificity This antibody is specific for isoform 1-1L.
Concentration 1mg/mL
Gene Accession No. P53805
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mL
Gene ID (Entrez) 1827
Cross Reactivity Human
Cross Reactivity Mouse
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Regulatory Status RUO
Antigen RCAN1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human RCAN1 (within residues 40-90). [Swiss-Prot No. P53805]