Please login to your online account to display your discounted pricing

anti-RBFOX3/NeuN, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human RBFOX3/NeuN protein (within residues 20-100). [Swiss-Prot A6NFN3]
Antigen RBFOX3/NeuN
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP177686

 
novus biologicals canada
NBP177686
Unlabeled 0.1mL Each for $362.00

NBP177686SS

 
novus biologicals canada
NBP177686SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human RBFOX3/NeuN protein (within residues 20-100). [Swiss-Prot A6NFN3]
Antigen RBFOX3/NeuN
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Test Specificity Human, rat and mouse. Immunogen has 95% identity to bovine.
Gene Accession No. A6NFN3
Gene ID (Entrez) 146713
Formulation PBS, 30% glycerol
Concentration 1.12mg/mL