Please login to your online account to display your discounted pricing

anti-RBFOX3/NeuN, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177686SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177686SS

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human, rat and mouse. Immunogen has 95% identity to bovine.
Conjugate Unlabeled
Gene Accession No. A6NFN3
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 0.025mL
Host Species Rabbit
Formulation PBS, 30% glycerol
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 146713
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Immunogen A synthetic peptide made to an internal region of the human RBFOX3/NeuN protein (within residues 20-100). [Swiss-Prot A6NFN3]
Concentration 1.12mg/mL
Regulatory Status RUO
Antigen RBFOX3/NeuN
Format Affinity Purified