Please login to your online account to display your discounted pricing

anti-RBFOX3/NeuN, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177686

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177686

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Format Affinity Purified
Gene Accession No. A6NFN3
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Primary or Secondary Primary
Antigen RBFOX3/NeuN
Host Species Rabbit
Gene ID (Entrez) 146713
Concentration 1.12mg/mL
Test Specificity Human, rat and mouse. Immunogen has 95% identity to bovine.
Immunogen A synthetic peptide made to an internal region of the human RBFOX3/NeuN protein (within residues 20-100). [Swiss-Prot A6NFN3]
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.1mL