Please login to your online account to display your discounted pricing

anti-RBCK1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 from human RBCk1 was used as the immunogen for this antibody.
Antigen RBCK1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227105

 
novus biologicals canada
NBP227105
Unlabeled 0.1mg Each for $255.80

NBP227105SS

 
novus biologicals canada
NBP227105SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 from human RBCk1 was used as the immunogen for this antibody.
Antigen RBCK1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, orangutan, and chimpanzee, 94% in rhesus monkey, 89% in new world monkey, and macaque, 83% in cow, guinea pig, and mole rat, 78% in dog, elephant, and rabbit, 72% in panda, 67% in opossum, 61% in rat, and 55% homologous in mouse.
Gene Accession No. Q9BYM8
Gene ID (Entrez) 10616
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL