Please login to your online account to display your discounted pricing

anti-RBBP6, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human RBBP6 protein (between residues 20-70) [UniProt Q7Z6E9]
Antigen RBBP6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP149542SS

 
novus biologicals canada
NBP149542SS
Unlabeled 0.025mL Each for $109.00

NBP149542

 
novus biologicals canada
NBP149542
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human RBBP6 protein (between residues 20-70) [UniProt Q7Z6E9]
Antigen RBBP6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Human. Predicted to react with mouse based on 100% sequence homology.
Gene Accession No. Q7Z6E9
Gene ID (Entrez) 5930
Formulation PBS, 30% glycerol
Concentration 1mg/mL