Please login to your online account to display your discounted pricing

anti-RBBP6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149542

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149542

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene Accession No. Q7Z6E9
Test Specificity Human. Predicted to react with mouse based on 100% sequence homology.
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Primary or Secondary Primary
Gene ID (Entrez) 5930
Concentration 1mg/mL
Cross Reactivity Human
Immunogen A synthetic peptide made to an internal portion of the human RBBP6 protein (between residues 20-70) [UniProt Q7Z6E9]
Host Species Rabbit
Format Affinity Purified
Antigen RBBP6
Regulatory Status RUO