Please login to your online account to display your discounted pricing

anti-rba-1, Polyclonal, Novus Biologicals™

$583.17 - $583.17

Specifications

Host Species Rabbit
Immunogen In vivo generated recominant protein fragment
Antigen rba-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

48750002

 
novus biologicals canada
48750002
Unlabeled 0.05mL Each for $583.17
Description & Specifications

Specifications

Host Species Rabbit
Immunogen In vivo generated recominant protein fragment
Antigen rba-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Test Specificity Caenorhabditis elegans.
Gene ID (Entrez) 172801
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
Concentration 1mg/mL