missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-RB1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 251-268 (RTPRRGQNRSARIAKQLE) of human Rb was used as immunogen
Antigen RB1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056888 Novus Biologicals Canada
NB10056888
0.2mL Each for $255.80
NB10056888T Novus Biologicals Canada
NB10056888SS
0.05mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
RB1
Purified
RUO
In Jurkat, a 110 kDa band is observed.
5925
0.5mg/mL
A synthetic peptide corresponding to amino acids 251-268 (RTPRRGQNRSARIAKQLE) of human Rb was used as immunogen
Polyclonal
Primary
Western Blot
P06400
PBS containing 0.2% gelatin