Please login to your online account to display your discounted pricing

anti-TLR9, Clone: 2A4C6.2E5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231150

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP231150

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype κ
Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Conjugate Unlabeled
Immunogen Partial recombinant protein made to an internal portion of human TLR9 (between amino acids 500-800) [UniProt Q9NR96]
Antigen TLR9
Format Purified
Host Species Rat
Regulatory Status RUO
Quantity 0.1mg
Primary or Secondary Primary
Clone 2A4C6.2E5
Cross Reactivity Mouse
Formulation PBS
Gene Accession No. Q9NR96
Gene ID (Entrez) 54106