Please login to your online account to display your discounted pricing

anti-TLR9, Clone: 2A4C6.2E5, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 2A4C6.2E5
Host Species Rat
Isotype κ
Immunogen Partial recombinant protein made to an internal portion of human TLR9 (between amino acids 500-800) [UniProt Q9NR96]
Antigen TLR9
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP231150

 
novus biologicals canada
NBP231150
Unlabeled 0.1mg Each for $309.05

NBP231150SS

 
novus biologicals canada
NBP231150SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 2A4C6.2E5
Host Species Rat
Isotype κ
Immunogen Partial recombinant protein made to an internal portion of human TLR9 (between amino acids 500-800) [UniProt Q9NR96]
Antigen TLR9
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. Q9NR96
Gene ID (Entrez) 54106
Formulation PBS
Concentration 1.0mg/mL