Please login to your online account to display your discounted pricing

anti-TLR9, Clone: 2A4C6.2E5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231150SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP231150SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 54106
Isotype κ
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Mouse
Quantity 0.025mg
Immunogen Partial recombinant protein made to an internal portion of human TLR9 (between amino acids 500-800) [UniProt Q9NR96]
Format Purified
Host Species Rat
Antigen TLR9
Clone 2A4C6.2E5
Concentration 1.0mg/mL
Monoclonal or Polyclonal Monoclonal
Gene Accession No. Q9NR96
Formulation PBS
Conjugate Unlabeled