Please login to your online account to display your discounted pricing

anti-TLR8, Biotin, Clone: 103E11.01, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0482B

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0482B

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 51311
Quantity 0.1mg
Isotype IgG2a
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Format Ion Exchange Chromatography
Conjugate Biotin
Formulation Antibody in PBS 50% glycerol
Primary or Secondary Primary
Host Species Rat
Antigen TLR8
Immunogen Human recombinant TLR8
Clone 103E11.01
Monoclonal or Polyclonal Monoclonal