Please login to your online account to display your discounted pricing

anti-TLR8, Biotin, Clone: 103E11.01, Novus Biologicals™

$406.00 - $536.00

Specifications

Clone 103E11.01
Host Species Rat
Isotype IgG2a
Immunogen Human recombinant TLR8
Antigen TLR8
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX0482B

 
novus biologicals canada
DDX0482B
Biotin 0.1mg Each for $536.00

DDX0482B005

 
novus biologicals canada
DDX0482B005MG
Biotin 0.05mg Each for $406.00
Description & Specifications

Specifications

Clone 103E11.01
Host Species Rat
Isotype IgG2a
Immunogen Human recombinant TLR8
Antigen TLR8
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene ID (Entrez) 51311
Formulation Antibody in PBS 50% glycerol