Please login to your online account to display your discounted pricing

anti-TLR8, Biotin, Clone: 103E11.01, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0482B005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0482B005

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 51311
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Human recombinant TLR8
Antigen TLR8
Regulatory Status RUO
Format Ion Exchange Chromatography
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rat
Formulation Antibody in PBS 50% glycerol
Primary or Secondary Primary
Conjugate Biotin
Monoclonal or Polyclonal Monoclonal
Quantity 0.05mg
Isotype IgG2a
Clone 103E11.01