Please login to your online account to display your discounted pricing

anti-TLR3, Clone: 27N3D4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227373

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227373

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Isotype κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7098
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Quantity 0.1mg
Antigen TLR3
Immunogen A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody.
Applications Flow Cytometry
Conjugate Unlabeled
Regulatory Status RUO
Host Species Rat
Clone 27N3D4