Please login to your online account to display your discounted pricing

anti-TLR3, Clone: 27N3D4, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 27N3D4
Host Species Rat
Isotype κ
Immunogen A portion of amino acids 340-390 of mouse TLR3 was used as the immunogen for this antibody.
Antigen TLR3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227404

 
novus biologicals canada
NBP227404
Unlabeled 0.1mg Each for $307.00

NBP227404SS

 
novus biologicals canada
NBP227404SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 27N3D4
Host Species Rat
Isotype κ
Immunogen A portion of amino acids 340-390 of mouse TLR3 was used as the immunogen for this antibody.
Antigen TLR3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blotting
Gene ID (Entrez) 7098
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL