Please login to your online account to display your discounted pricing

anti-TLR3, Clone: 27N3D4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227373SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227373SS

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Conjugate Unlabeled
Primary or Secondary Primary
Quantity 0.025mg
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Regulatory Status RUO
Clone 27N3D4
Cross Reactivity Human
Cross Reactivity Mouse
Isotype κ
Host Species Rat
Format Purified
Gene ID (Entrez) 7098
Applications Flow Cytometry
Immunogen A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody.
Antigen TLR3