Please login to your online account to display your discounted pricing

anti-TLR2, Clone: 11G5, Novus Biologicals™

$139.00 - $259.02

Specifications

Clone 11G5
Host Species Rat
Isotype κ
Immunogen A portion of amino acids 400-450 of murine TLR2 was used as the immunogen for this antibody.
Antigen TLR2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227165

 
novus biologicals canada
NBP227165
Unlabeled 0.1mg Each for $259.02

NBP227165SS

 
novus biologicals canada
NBP227165SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 11G5
Host Species Rat
Isotype κ
Immunogen A portion of amino acids 400-450 of murine TLR2 was used as the immunogen for this antibody.
Antigen TLR2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 85% homologous in Vole, 80% in Rat, Chinese Hamster and Rabbit, 70% in Orangutan, Rhesus Monkey and Horse, and 65% homologous in Human.
Gene ID (Entrez) 7097
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL