Please login to your online account to display your discounted pricing

anti-TfR (Transferrin R), Clone: 8D3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10064979

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10064979

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications In vivo assay
Antigen TfR (Transferrin R)
Immunogen Mouse transformed endothelioma cell line t.end1.
Isotype IgG2a
Cross Reactivity Mouse
Clone 8D3
Regulatory Status RUO
Quantity 0.25mg
Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Formulation PBS, pH 7.4
Gene ID (Entrez) 7037
Primary or Secondary Primary
Conjugate Unlabeled
Host Species Rat
Format Purified
Test Specificity NB100-64979 recognizes native, soluble and denatured forms of murine CD71. The antibody has been used as a BBB transporter vector in mice and is suitable for studying CD71 expression, and iron uptake into different tissues, in the mouse.