Please login to your online account to display your discounted pricing

anti-Podoplanin, Clone: pmab-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203955

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203955

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10630
Concentration 0.5mg/mL
Quantity 0.1mg
Isotype κ
Cross Reactivity Mouse
Format Purified
Immunogen The immunogen is Podoplanin.
Host Species Rat
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blotting
Antigen Podoplanin
Clone pmab-1
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal