Please login to your online account to display your discounted pricing

anti-Podoplanin, Clone: pmab-1, Novus Biologicals™

$109.00 - $219.00

Specifications

Clone pmab-1
Host Species Rat
Isotype κ
Immunogen The immunogen is Podoplanin.
Antigen Podoplanin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203955SS

 
novus biologicals canada
NBP203955SS
Unlabeled 0.025mg Each for $109.00

NBP203955

 
novus biologicals canada
NBP203955
Unlabeled 0.1mg Each for $219.00
Description & Specifications

Specifications

Clone pmab-1
Host Species Rat
Isotype κ
Immunogen The immunogen is Podoplanin.
Antigen Podoplanin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blot
Gene ID (Entrez) 10630
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL