Please login to your online account to display your discounted pricing

anti-Podoplanin, Clone: pmab-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203955SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203955SS

  • / Each
 

Description & Specifications

Specifications

Format Purified
Immunogen The immunogen is Podoplanin.
Isotype κ
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse
Antigen Podoplanin
Host Species Rat
Quantity 0.025mg
Applications Flow Cytometry
Applications Western Blotting
Clone pmab-1
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Gene ID (Entrez) 10630
Concentration 0.5mg/mL