Please login to your online account to display your discounted pricing

anti-Oval Cell Marker, Clone: OC2-2A6, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone OC2-2A6
Host Species Rat
Isotype IgG1
Immunogen Nonparenchymal cells from DDC-treated mice to induce the oval cell response.
Antigen Oval Cell Marker
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP118967SS

 
novus biologicals canada
NBP118967SS
Unlabeled 0.025mL Each for $109.00

NBP118967

 
novus biologicals canada
NBP118967
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone OC2-2A6
Host Species Rat
Isotype IgG1
Immunogen Nonparenchymal cells from DDC-treated mice to induce the oval cell response.
Antigen Oval Cell Marker
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Test Specificity Specific for proliferating hepatic periductal cells.
Concentration 1mg/mL