Please login to your online account to display your discounted pricing

anti-Noxa, Clone: 69N15G8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225252

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225252

  • / Each
 

Description & Specifications

Specifications

Format Purified
Isotype κ
Cross Reactivity Mouse
Host Species Rat
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Quantity 0.1mg
Antigen Noxa
Regulatory Status RUO
Applications Western Blotting
Clone 69N15G8
Test Specificity This product reacts with Mouse.
Conjugate Unlabeled
Formulation Noxa antibody at 0.5mg/mL in PBS containing, 0.05% BSA
Concentration 0.5mg/mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5366