Please login to your online account to display your discounted pricing

anti-Noxa, Clone: 69N15G8, Novus Biologicals™

$109.00 - $252.00

Specifications

Clone 69N15G8
Host Species Rat
Isotype κ
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Antigen Noxa
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225252

 
novus biologicals canada
NBP225252
Unlabeled 0.1mg Each for $252.00

NBP225252SS

 
novus biologicals canada
NBP225252SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 69N15G8
Host Species Rat
Isotype κ
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Antigen Noxa
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This product reacts with Mouse.
Gene ID (Entrez) 5366
Formulation Noxa antibody at 0.5mg/mL in PBS containing, 0.05% BSA
Concentration 0.5mg/mL