Please login to your online account to display your discounted pricing

anti-Noxa, Clone: 69N15G8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225267SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225267SS

  • / Each
 

Description & Specifications

Specifications

Antigen Noxa
Isotype κ
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Format Purified
Applications Western Blotting
Test Specificity This product reacts with Mouse.
Concentration 0.5mg/mL
Primary or Secondary Primary
Formulation Noxa antibody at 0.5mg/mL in sterile PBS
Monoclonal or Polyclonal Monoclonal
Quantity 0.025mg
Gene ID (Entrez) 5366
Conjugate Unlabeled
Host Species Rat
Regulatory Status RUO
Cross Reactivity Mouse
Clone 69N15G8