Please login to your online account to display your discounted pricing

anti-Noxa, Clone: 69N15G8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225267

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225267

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Cross Reactivity Mouse
Clone 69N15G8
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5366
Concentration 0.5mg/mL
Formulation Noxa antibody at 0.5mg/mL in sterile PBS
Test Specificity This product reacts with Mouse.
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Conjugate Unlabeled
Antigen Noxa
Format Purified
Host Species Rat