Please login to your online account to display your discounted pricing

anti-Noxa, Clone: 69N15G8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225252SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225252SS

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Quantity 0.025mg
Test Specificity This product reacts with Mouse.
Cross Reactivity Mouse
Format Purified
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Host Species Rat
Applications Western Blotting
Regulatory Status RUO
Antigen Noxa
Clone 69N15G8
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 5366
Formulation Noxa antibody at 0.5mg/mL in PBS containing, 0.05% BSA
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal